Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 29 - 30 | 4593 | 94 | |
GenBank mRNA | AB299229.1 | - | 29 - 30 | 4520 | 94 | |
AK046668.1 | - | 8 - 9 | 1082 | 94 | ||
BC047418.1 | - | 19 - 20 | 2706 | 94 | ||
BC094568.1 | - | 11 - 12 | 1877 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
