Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 7 - 8 | 1292 | 108 | |
GenBank mRNA | AB299229.1 | - | 7 - 8 | 1219 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
