Assay Details
Gene Symbol: Scyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198021.2 | NP_932138.1 | 4 - 5 | 749 | 148 | |
GenBank mRNA | AK139761.1 | - | 4 - 5 | 733 | 148 | |
BC042443.1 | - | 4 - 5 | 749 | 4037878 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
