Assay Details
Gene Symbol: Gm4710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033456.1 | 2 - 3 | 403 | 97 | ||
GenBank mRNA | AK082440.1 | - | 2 - 3 | 404 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
family with sequence similarity 98, member A |
Fam98a |
2810405J04Rik, AL024114 |
-309.6kb | Anti-sense | Chr. 17 - 75537086 - 75551946 on NCBI Build GRCm38 |
Downstream |
predicted gene, 32282 |
Gm32282 | 2.3mb | Anti-sense | Chr. 17 - 78199250 - 78200084 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663933 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139968 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231843 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740250 | AK042157.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | AK054056.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | AK088875.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | AK154893.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | BC028512.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | BC131655.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | M11934.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | M14342.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | M14343.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | M92933.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | NM_001111316.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385124 | NM_011210.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880751 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198740 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77421 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112267 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138009 | NR_033456.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138009 | AK082440.1 | mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359376 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359263 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13438392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13143849 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359248 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13361825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359321 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359243 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359375 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359245 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359255 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13186317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359442 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359238 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359385 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359378 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13087961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359381 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13185934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359465 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13087951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359468 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13317198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13092676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359252 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13312265 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13181737 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13375075 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359429 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359260 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13144604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13144602 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13144606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359440 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359422 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359323 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359259 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359269 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359476 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13247582 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359258 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359231 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359372 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13496083 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359251 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359329 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13131449 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13139736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359247 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13271241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359417 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359240 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13092683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31359472 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1700011169.mm.1 | TC1700001174.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700011169.mm.1 | NR_033456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011169.mm.1 | NR_033456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011170.mm.1 | TC1700001174.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700011170.mm.1 | NR_033456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011170.mm.1 | NR_033456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011171.mm.1 | TC1700001174.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700011171.mm.1 | NR_033456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011171.mm.1 | NR_033456.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 533941 | - | - |
