Assay Details
Gene Symbol: Sema4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090073.1 | - | 7 - 8 | 878 | 86 | |
AK148298.1 | - | 7 - 8 | 878 | 86 | ||
AK173247.1 | - | 8 - 9 | 1118 | 86 | ||
S79463.1 | - | 7 - 8 | 732 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
