Assay Details
Gene Symbol: Mst1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009074.2 | NP_033100.2 | 8 - 9 | 2603 | 75 | |
NR_109782.1 | 8 - 9 | 2631 | 75 | |||
GenBank mRNA | AK053655.1 | - | 8 - 9 | 2614 | 75 | |
BC141379.1 | - | 8 - 9 | 2414 | 9055992 | 75 | |
X74736.1 | - | 8 - 9 | 2603 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
