Assay Details
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011842.3 | NP_035972.3 | 14 - 15 | 1919 | 68 | |
GenBank mRNA | AF159259.1 | - | 14 - 15 | 1794 | 68 | |
AK003701.1 | - | 2 - 3 | 353 | 68 | ||
AK077548.1 | - | 14 - 15 | 1901 | 68 | ||
AK147099.1 | - | 14 - 15 | 1898 | 68 | ||
AK149834.1 | - | 14 - 15 | 1965 | 68 | ||
AK157717.1 | - | 14 - 15 | 1823 | 68 | ||
AK159979.1 | - | 14 - 15 | 1895 | 68 | ||
AK185607.1 | - | 1 - 2 | 158 | 68 | ||
AK218258.1 | - | 1 - 2 | 158 | 68 | ||
BC079847.1 | - | 14 - 15 | 1919 | 6419411 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
