Assay Details
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011842.3 | NP_035972.3 | 4 - 5 | 746 | 139 | |
GenBank mRNA | AF159259.1 | - | 4 - 5 | 621 | 139 | |
AK077548.1 | - | 4 - 5 | 728 | 139 | ||
AK088158.1 | - | 4 - 5 | 659 | 139 | ||
AK147099.1 | - | 4 - 5 | 725 | 139 | ||
AK149834.1 | - | 4 - 5 | 792 | 139 | ||
AK157717.1 | - | 4 - 5 | 650 | 139 | ||
AK159979.1 | - | 4 - 5 | 722 | 139 | ||
AK180159.1 | - | 2 - 3 | 173 | 139 | ||
BC079847.1 | - | 4 - 5 | 746 | 6419411 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
