Assay Details
Gene Symbol: Hnrnpul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081196.1 | NP_001074665.1 | 2 - 3 | 1550 | 86 | |
GenBank mRNA | DQ000354.1 | - | 2 - 3 | 669 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
