Assay Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164634.1 | NP_001158106.1 | 3 - 4 | 666 | 67 | |
NM_001164635.1 | NP_001158107.1 | 3 - 4 | 574 | 67 | ||
NM_031194.5 | NP_112471.3 | 4 - 5 | 746 | 67 | ||
GenBank mRNA | AB079895.1 | - | 2 - 3 | 403 | 67 | |
AF078869.1 | - | 3 - 4 | 521 | 67 | ||
AK031962.1 | - | 4 - 5 | 712 | 67 | ||
AK044336.1 | - | 3 - 4 | 543 | 67 | ||
AK157891.1 | - | 3 - 4 | 575 | 67 | ||
BC014762.1 | - | 3 - 4 | 552 | 4239544 | 67 | |
BU615016.1 | - | 3 - 4 | 616 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
