Assay Details
Gene Symbol: Tcirg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136091.2 | NP_001129563.1 | 9 - 10 | 1138 | 68 | |
NM_001167784.1 | NP_001161256.1 | 9 - 10 | 1483 | 68 | ||
NM_016921.3 | NP_058617.3 | 9 - 10 | 1179 | 68 | ||
GenBank mRNA | AB022322.1 | - | 9 - 10 | 1045 | 68 | |
AF218253.1 | - | 9 - 10 | 1117 | 68 | ||
AK155140.1 | - | 9 - 10 | 1484 | 68 | ||
BC006761.1 | - | 9 - 10 | 1179 | 3257437 | 68 | |
BC085234.1 | - | 9 - 10 | 1039 | 5322429 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
