Assay Details
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017792.1 | - | 2 - 3 | 287 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 7656 | Mir7656 |
Gm27614, mmu-mir-7656 |
-370.1kb | Sense | Chr. 9 - 94751829 - 94751890 on NCBI Build GRCm38 |
Downstream | carbohydrate sulfotransferase 2 | Chst2 |
AI428561, AW121776, C130041E03Rik, Chts2, GST-2, GlcNAc6ST, Gn6st |
276.4kb | Anti-sense | Chr. 9 - 95400926 - 95407270 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
