Assay Details
Gene Symbol: Mta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171052.1 | NP_001164523.1 | 2 - 3 | 197 | 97 | |
NM_001171053.1 | NP_001164524.1 | 2 - 3 | 197 | 97 | ||
NM_001171054.1 | NP_001164525.1 | 2 - 3 | 197 | 97 | ||
NM_054082.2 | NP_473423.1 | 2 - 3 | 197 | 97 | ||
GenBank mRNA | AF288138.1 | - | 2 - 3 | 151 | 97 | |
AK129318.1 | - | 2 - 3 | 126 | 97 | ||
AK145942.1 | - | 2 - 3 | 142 | 97 | ||
AK146721.1 | - | 2 - 3 | 150 | 97 | ||
AK146905.1 | - | 2 - 3 | 150 | 97 | ||
AK147197.1 | - | 2 - 3 | 140 | 97 | ||
AK154856.1 | - | 2 - 3 | 137 | 97 | ||
BC022124.1 | - | 2 - 3 | 161 | 5097283 | 97 | |
BC099960.1 | - | 2 - 3 | 114 | 30023887 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
