Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024930.mm.1 |
TC0900003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024930.mm.1 |
NM_198931.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024930.mm.1 |
NM_198931.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024932.mm.1 |
TC0900003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024932.mm.1 |
NM_026447.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024932.mm.1 |
NM_026447.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024926.mm.1 |
TC0900003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024926.mm.1 |
NM_026447.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024926.mm.1 |
NM_026447.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024926.mm.1 |
NM_198931.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024926.mm.1 |
NM_198931.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024924.mm.1 |
TC0900003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024924.mm.1 |
NM_026447.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024924.mm.1 |
NM_026447.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024924.mm.1 |
NM_198931.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024924.mm.1 |
NM_198931.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024928.mm.1 |
TC0900003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024928.mm.1 |
NM_026447.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024928.mm.1 |
NM_026447.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024928.mm.1 |
NM_198931.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024928.mm.1 |
NM_198931.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024927.mm.1 |
TC0900003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024927.mm.1 |
NM_026447.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024927.mm.1 |
NM_026447.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024927.mm.1 |
NM_198931.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024927.mm.1 |
NM_198931.3 |
Refseq |