Assay Details
Gene Symbol: Ppm1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026447.4 | NP_080723.3 | 9 - 10 | 1285 | 61 | |
NM_198931.3 | NP_945149.2 | 9 - 10 | 1289 | 61 | ||
GenBank mRNA | AB474373.1 | - | 9 - 10 | 1324 | 61 | |
AK013149.1 | - | 9 - 10 | 1218 | 61 | ||
AK017245.1 | - | 6 - 7 | 622 | 61 | ||
AK046387.1 | - | 1184 | 61 | |||
AK087999.1 | - | 9 - 10 | 1323 | 61 | ||
AY332616.1 | - | 9 - 10 | 1292 | 61 | ||
BC058248.1 | - | 9 - 10 | 1211 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801800 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801799 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801802 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801825 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801801 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801798 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033510 | BC058248.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033510 | AK046387.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033510 | AK087999.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033510 | NM_198931.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033510 | AB474373.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138075 | AK046387.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149264 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726201 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706863 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118099 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033510 | BC058248.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033510 | AK046387.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033510 | AK087999.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033510 | NM_198931.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033510 | AB474373.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P252545 | BC058248.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P252545 | AK046387.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P252545 | NM_198931.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P252545 | AK013149.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P252545 | AK017245.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801813 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801788 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801793 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801790 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801820 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801818 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801800 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801795 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801823 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801825 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801815 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801777 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801796 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801786 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801810 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26801806 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118099 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06914261 | - | - |
Whole Mouse Genome G4122A | A_51_P252545 | BC058248.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P252545 | AK046387.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P252545 | NM_198931.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P252545 | AK013149.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P252545 | AK017245.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130499 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118099 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07672950 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08110684 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07672962 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08110686 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08098874 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07672953 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08098873 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07672963 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07672948 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801799 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801825 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801798 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801801 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801802 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26801800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26801784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07524539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801808 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07524534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07524548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07524516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07672950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07672962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07995542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07672961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801810 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26801757 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10596445 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596452 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596449 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596453 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596446 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596451 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596444 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596450 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596448 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596447 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428351_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10596442 | - | - |
Mouse Genome 430 2.0 Array | 1428351_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1428351_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67905_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67905_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900024916.mm.1 | TC0900003003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900024916.mm.1 | NM_026447.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024916.mm.1 | NM_026447.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024916.mm.1 | NM_198931.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024916.mm.1 | NM_198931.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024918.mm.1 | TC0900003003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900024918.mm.1 | NM_026447.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024918.mm.1 | NM_026447.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024918.mm.1 | NM_198931.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024918.mm.1 | NM_198931.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024917.mm.1 | TC0900003003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900024917.mm.1 | NM_026447.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024917.mm.1 | NM_026447.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024917.mm.1 | NM_198931.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024917.mm.1 | NM_198931.3 | Refseq |
Mouse Expression Set 430 | 1428351_PM_at | - | - |
Mouse Expression Set 430 | 1428351_at | - | - |
Murine Genome U74v2 Set | 104525_PM_at | - | - |
Murine Genome U74v2 Set | 161992_at | - | - |
Murine Genome U74v2 Set | 161992_PM_at | - | - |
Murine Genome U74v2 Set | 104525_at | - | - |
Mouse Genome 430A 2.0 Array | 1428351_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1428351_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428351_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428351_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5270279 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3780577 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5270279 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005270279 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003780577 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224437 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251484 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224437 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1251484 | - | - |
mouse-6 v1.1 (Illumina) | scl067905.1_18-S | - | - |
mouse-6 v1.1 (Illumina) | scl0003494.1_23-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5270279 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3780577 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067905.1_18-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003494.1_23-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270279 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780577 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3151840 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1251484 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3073044 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251484 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3073044 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224437 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3151840 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067905.1_18-S | - | - |
