Assay Details
Gene Symbol: Ranbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252466.1 | NP_001239395.1 | 7 - 8 | 711 | 120 | |
NM_027933.3 | NP_082209.1 | 6 - 7 | 510 | 120 | ||
NR_045519.1 | 7 - 8 | 620 | 120 | |||
GenBank mRNA | AK040481.1 | - | 8 - 9 | 694 | 120 | |
AK133949.1 | - | 6 - 7 | 357 | 120 | ||
AK155301.1 | - | 6 - 7 | 379 | 120 | ||
BC066005.1 | - | 6 - 7 | 351 | 120 | ||
BC145892.1 | - | 6 - 7 | 374 | 40131195 | 120 | |
BC145894.1 | - | 6 - 7 | 374 | 40131197 | 120 | |
BM950080.1 | - | 6 - 7 | 445 | 120 | ||
CA318306.1 | - | 7 - 8 | 546 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
