Assay Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042513.1 | NP_001035978.1 | 10 - 11 | 1176 | 70 | |
NM_001042514.1 | NP_001035979.1 | 9 - 10 | 1264 | 70 | ||
NM_001042523.1 | NP_001035988.1 | 10 - 11 | 1393 | 70 | ||
NM_015762.2 | NP_056577.2 | 9 - 10 | 1069 | 70 | ||
GenBank mRNA | AB027565.1 | - | 9 - 10 | 1096 | 70 | |
AF333036.1 | - | 10 - 11 | 1393 | 70 | ||
AK146125.1 | - | 9 - 10 | 1070 | 70 | ||
AK149625.1 | - | 10 - 11 | 1177 | 70 | ||
AK168356.1 | - | 10 - 11 | 1177 | 70 | ||
BC020274.1 | - | 9 - 10 | 1031 | 70 | ||
BC037643.1 | - | 9 - 10 | 1349 | 5059831 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
