Assay Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042513.1 | NP_001035978.1 | 3 - 4 | 331 | 112 | |
NM_001042514.1 | NP_001035979.1 | 2 - 3 | 419 | 112 | ||
NM_001042523.1 | NP_001035988.1 | 3 - 4 | 548 | 112 | ||
NM_015762.2 | NP_056577.2 | 2 - 3 | 224 | 112 | ||
GenBank mRNA | AB027565.1 | - | 2 - 3 | 251 | 112 | |
AF333036.1 | - | 3 - 4 | 548 | 112 | ||
AK011902.1 | - | 3 - 4 | 332 | 112 | ||
AK146125.1 | - | 2 - 3 | 225 | 112 | ||
AK149625.1 | - | 3 - 4 | 332 | 112 | ||
AK151927.1 | - | 3 - 4 | 332 | 112 | ||
AK168356.1 | - | 3 - 4 | 332 | 112 | ||
AK172383.1 | - | 2 - 3 | 257 | 112 | ||
AK177272.1 | - | 2 - 3 | 151 | 112 | ||
AK186760.1 | - | 2 - 3 | 171 | 112 | ||
AK194169.1 | - | 2 - 3 | 169 | 112 | ||
AK210892.1 | - | 2 - 3 | 151 | 112 | ||
BC020274.1 | - | 2 - 3 | 186 | 112 | ||
BC037643.1 | - | 2 - 3 | 504 | 5059831 | 112 | |
CJ136679.1 | - | 3 - 4 | 332 | 112 | ||
CX562215.1 | - | 2 - 3 | 419 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
