Assay Details
Gene Symbol: Adgre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010130.4 | NP_034260.1 | 10 - 11 | 1229 | 115 | |
GenBank mRNA | AK133202.1 | - | 10 - 11 | 1205 | 115 | |
AK149898.1 | - | 10 - 11 | 1215 | 115 | ||
AK151303.1 | - | 10 - 11 | 1201 | 115 | ||
AK151682.1 | - | 10 - 11 | 1221 | 115 | ||
AK155862.1 | - | 10 - 11 | 1180 | 115 | ||
BC075688.1 | - | 10 - 11 | 1201 | 30609706 | 115 | |
U66888.1 | - | 10 - 11 | 1216 | 115 | ||
X93328.1 | - | 10 - 11 | 1225 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
