Assay Details
Gene Symbol: Ptprr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011217.2 | NP_035347.1 | 1 - 2 | 451 | 93 | |
GenBank mRNA | AK028054.1 | - | 1 - 2 | 452 | 93 | |
AK051719.1 | - | 1 - 2 | 528 | 93 | ||
BC119230.1 | - | 1 - 2 | 384 | 8733979 | 93 | |
BC120547.1 | - | 1 - 2 | 384 | 40129470 | 93 | |
D31898.1 | - | 1 - 2 | 414 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
