Assay Details
Gene Symbol: Sema3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011348.2 | NP_035478.2 | 10 - 11 | 1781 | 92 | |
GenBank mRNA | AF034744.1 | - | 10 - 11 | 1516 | 92 | |
AK129116.1 | - | 5 - 6 | 546 | 92 | ||
AK141295.1 | - | 9 - 10 | 2503 | 92 | ||
AK142340.1 | - | 10 - 11 | 1608 | 92 | ||
AK159214.1 | - | 10 - 11 | 1572 | 92 | ||
BC057956.1 | - | 10 - 11 | 1679 | 5357516 | 92 | |
Z80941.1 | - | 10 - 11 | 1147 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
