Assay Details
Gene Symbol: Sema3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011348.2 | NP_035478.2 | 1 - 2 | 746 | 116 | |
GenBank mRNA | AF034744.1 | - | 1 - 2 | 481 | 116 | |
AK141295.1 | - | 1 - 2 | 748 | 116 | ||
AK142340.1 | - | 1 - 2 | 573 | 116 | ||
AK159214.1 | - | 1 - 2 | 537 | 116 | ||
BC057956.1 | - | 1 - 2 | 644 | 5357516 | 116 | |
Z80941.1 | - | 1 - 2 | 112 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
