Assay Details
Gene Symbol: Lmbrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK032560.1 | - | 574 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK032560.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
