Assay Details
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008928.4 | NP_032954.1 | 8 - 9 | 866 | 77 | |
GenBank mRNA | AK008141.1 | - | 8 - 9 | 869 | 77 | |
AK011002.1 | - | 8 - 9 | 867 | 77 | ||
AK053378.1 | - | 8 - 9 | 867 | 77 | ||
AK150067.1 | - | 8 - 9 | 867 | 77 | ||
AK159379.1 | - | 8 - 9 | 865 | 77 | ||
AK166255.1 | - | 8 - 9 | 868 | 77 | ||
AK166507.1 | - | 8 - 9 | 867 | 77 | ||
AK167209.1 | - | 8 - 9 | 867 | 77 | ||
AK167442.1 | - | 8 - 9 | 870 | 77 | ||
AK168710.1 | - | 8 - 9 | 849 | 77 | ||
BC007467.1 | - | 7 - 8 | 805 | 3497714 | 77 | |
D87115.1 | - | 8 - 9 | 879 | 77 | ||
X93150.1 | - | 6 - 7 | 601 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
