Assay Details
Gene Symbol: Gm14858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040285.1 | 2 - 3 | 509 | 67 | ||
GenBank mRNA | AK090266.1 | - | 2 - 3 | 510 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1 | Cited1 |
AI316840, AU019144, Msg1 |
-1.3kb | Anti-sense | Chr. X - 102247378 - 102252181 on NCBI Build GRCm38 |
Downstream | histone deacetylase 8 | Hdac8 | 2610007D20Rik | 27.0kb | Anti-sense | Chr. X - 102284640 - 102509686 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
