Assay Details
Gene Symbol: Gm34184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035016.1 | - | 3 - 4 | 381 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1500009L16 gene | 1500009L16Rik |
2700050C19Rik, OCC-1 |
-280.8kb | Anti-sense | Chr. 10 - 83722666 - 83762762 on NCBI Build GRCm38 |
Downstream |
NUAK family, SNF1-like kinase, 1 |
Nuak1 |
AU014801, AW494241, B230104P22Rik, Omphk1 |
316.4kb | Sense | Chr. 10 - 84370900 - 84440471 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
