Assay Details
Gene Symbol: Gm12132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK042535.1 | - | 7 - 8 | 765 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930553C11 gene | 4930553C11Rik | -946.7kb | Anti-sense | Chr. 11 - 39307703 - 39390225 on NCBI Build GRCm38 | |
Downstream |
methionine adenosyltransferase II, beta |
Mat2b |
1110064C04Rik, 2410018D16Rik, AI182287, AU022853, MAT-II, MATIIbeta, TGR |
341.5kb | Anti-sense | Chr. 11 - 40679314 - 40695203 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
