Assay Details
Gene Symbol: C330024C12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046016.1 | 1 - 2 | 677 | 110 | ||
GenBank mRNA | AK082808.1 | - | 1 - 2 | 678 | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | suppressor of cytokine signaling 5 | Socs5 |
1810018L08Rik, Cish5, mKIAA0671 |
-55.3kb | Anti-sense | Chr. 17 - 87107407 - 87137647 on NCBI Build GRCm38 |
Downstream | multiple coagulation factor deficiency 2 | Mcfd2 |
1810021C21Rik, F5f8d, Lman1ip, Sdnsf |
59.5kb | Sense | Chr. 17 - 87254443 - 87265947 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
