Assay Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175236.4 | NP_780445.1 | 7 - 8 | 672 | 135 | |
NR_027664.1 | 6 - 7 | 631 | 135 | |||
GenBank mRNA | AK018241.1 | - | 7 - 8 | 646 | 135 | |
AK038853.1 | - | 7 - 8 | 639 | 135 | ||
AK046780.1 | - | 7 - 8 | 598 | 135 | ||
AK050178.1 | - | 6 - 7 | 596 | 135 | ||
AK084104.1 | - | 600 | 135 | |||
BC026584.1 | - | 7 - 8 | 630 | 4972323 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
