Assay Details
Gene Symbol: Gabrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008070.3 | NP_032096.1 | 1 - 2 | 278 | 95 | |
GenBank mRNA | AK090279.1 | - | 1 - 2 | 279 | 95 | |
BC145973.1 | - | 1 - 2 | 252 | 40131276 | 95 | |
BC145975.1 | - | 1 - 2 | 252 | 40131278 | 95 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK090279.1, BC145973.1, BC145975.1, NM_008070.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
