Assay Details
Gene Symbol: Gabrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008070.3 | NP_032096.1 | 5 - 6 | 882 | 72 | |
GenBank mRNA | AK034069.1 | - | 4 - 5 | 497 | 72 | |
AK090279.1 | - | 5 - 6 | 883 | 72 | ||
BC145973.1 | - | 5 - 6 | 856 | 40131276 | 72 | |
BC145975.1 | - | 5 - 6 | 856 | 40131278 | 72 | |
GU086167.1 | - | 4 - 5 | 455 | 72 | ||
U14419.1 | - | 4 - 5 | 546 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
