Assay Details
Gene Symbol: Gabrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008070.3 | NP_032096.1 | 9 - 10 | 1503 | 65 | |
GenBank mRNA | AK090279.1 | - | 9 - 10 | 1504 | 65 | |
BC145973.1 | - | 9 - 10 | 1477 | 40131276 | 65 | |
BC145975.1 | - | 9 - 10 | 1477 | 40131278 | 65 | |
U14419.1 | - | 8 - 9 | 1167 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
