Assay Details
Gene Symbol: Ppp1r21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028658.4 | NP_082934.3 | 12 - 13 | 1360 | 85 | |
GenBank mRNA | AK028185.1 | - | 12 - 13 | 1224 | 85 | |
AK155094.1 | - | 12 - 13 | 1192 | 85 | ||
AK170254.1 | - | 12 - 13 | 1248 | 85 | ||
BC137673.1 | - | 12 - 13 | 1300 | 8860692 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
