Assay Details
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011790.4 | NP_035920.1 | 5 - 6 | 685 | 94 | |
GenBank mRNA | AF124664.1 | - | 4 - 5 | 512 | 94 | |
AJ130975.1 | - | 4 - 5 | 544 | 94 | ||
AK011094.1 | - | 5 - 6 | 689 | 94 | ||
AK045012.1 | - | 668 | 94 | |||
AK149936.1 | - | 5 - 6 | 665 | 94 | ||
AK165028.1 | - | 5 - 6 | 623 | 94 | ||
AK167104.1 | - | 5 - 6 | 642 | 94 | ||
AK168055.1 | - | 5 - 6 | 690 | 94 | ||
BC051998.1 | - | 5 - 6 | 685 | 6400287 | 94 | |
BC052422.1 | - | 5 - 6 | 685 | 6400287 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
