Assay Details
Gene Symbol: Rps6ka4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019924.2 | NP_064308.2 | 14 - 15 | 1881 | 135 | |
GenBank mRNA | AF074714.1 | - | 14 - 15 | 1853 | 135 | |
AK004412.1 | - | 4 - 5 | 487 | 136 | ||
AK089467.1 | - | 14 - 15 | 1873 | 135 | ||
AK154674.1 | - | 14 - 15 | 1880 | 135 | ||
BC012964.1 | - | 14 - 15 | 1891 | 4160873 | 135 | |
CF616720.1 | - | 2 - 3 | 258 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
