Assay Details
Gene Symbol: Rps6ka4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019924.2 | NP_064308.2 | 2 - 3 | 205 | 56 | |
GenBank mRNA | AF074714.1 | - | 2 - 3 | 177 | 56 | |
AK089467.1 | - | 2 - 3 | 197 | 56 | ||
AK154674.1 | - | 2 - 3 | 204 | 56 | ||
BC012964.1 | - | 2 - 3 | 215 | 4160873 | 56 | |
CJ065157.1 | - | 2 - 3 | 200 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
