Assay Details
Gene Symbol: Safb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029979.2 | NP_001025150.2 | 15 - 16 | 2471 | 61 | |
GenBank mRNA | BC027837.1 | - | 2 - 3 | 437 | 61 | |
BC028827.1 | - | 9 - 10 | 1185 | 61 | ||
BC050855.1 | - | 15 - 16 | 2405 | 61 | ||
BC057039.1 | - | 13 - 14 | 1601 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
