Assay Details
Gene Symbol: Safb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029979.2 | NP_001025150.2 | 4 - 5 | 665 | 130 | |
GenBank mRNA | AK147798.1 | - | 4 - 5 | 605 | 130 | |
AK172898.1 | - | 4 - 5 | 641 | 130 | ||
BC050855.1 | - | 4 - 5 | 599 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
