Assay Details
Gene Symbol: Fam150a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195732.1 | NP_001182661.1 | 4 - 5 | 389 | 170 | |
GenBank mRNA | AK087278.1 | - | 4 - 5 | 265 | 170 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P151240 | AK087278.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P151240 | NM_001195732.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P151240 | AK087278.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P151240 | NM_001195732.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74323 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312919 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855810 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944742 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146896 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229865 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699296 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741713 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802922 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42970 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P151240 | AK087278.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P151240 | NM_001195732.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P151240 | AK087278.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P151240 | NM_001195732.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04005346 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04005365 | - | - |
Whole Mouse Genome G4122A | A_52_P151240 | AK087278.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P151240 | NM_001195732.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P151240 | AK087278.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P151240 | NM_001195732.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012312 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01048640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00477036 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012311 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012456 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00723356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01143754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00958822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00891944 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00011831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00316964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00477049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00011826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00699924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00477042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00576400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00742761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012348 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01048643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012404 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012350 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01048641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012518 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00958823 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00435694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20012525 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10344678 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10344676 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10344677 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10344675 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10344674 | - | - |
Affymetrix Mouse Clariom S Array | PSR0100000183.mm.1 | NM_001195732.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100000183.mm.1 | NM_001195732.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100000183.mm.1 | TC0100000022.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 789842 | - | - |
