Assay Details
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK133647.1 | - | 2174 | 71 | ||
M37890.1 | - | 4 - 5 | 2113 | 71 | ||
S56585.1 | - | 4 - 5 | 2192 | 71 | ||
X53779.1 | - | 4 - 5 | 2145 | 71 | ||
X59411.1 | - | 4 - 5 | 2144 | 71 | ||
X59592.1 | - | 4 - 5 | 2233 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
