Assay Details
Gene Symbol: Esyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028731.5 | NP_083007.2 | 8 - 9 | 1050 | 73 | |
GenBank mRNA | AK044360.1 | - | 8 - 9 | 1052 | 73 | |
AK157352.1 | - | 8 - 9 | 1019 | 73 | ||
BC138937.1 | - | 8 - 9 | 959 | 8861959 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK044360.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
