Assay Details
Gene Symbol: Chst10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145142.2 | NP_660124.2 | 3 - 4 | 516 | 78 | |
GenBank mRNA | AF360543.1 | - | 3 - 4 | 348 | 78 | |
AK046415.1 | - | 3 - 4 | 368 | 78 | ||
AK050676.1 | - | 2 - 3 | 398 | 78 | ||
BC056956.1 | - | 3 - 4 | 445 | 78 | ||
BC132223.1 | - | 2 - 3 | 188 | 40130500 | 78 | |
BC132225.1 | - | 2 - 3 | 188 | 40130502 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
