Assay Details
Gene Symbol: Mycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010850.2 | NP_034980.2 | 1 | 881 | 67 | |
GenBank mRNA | AK132789.1 | - | 1 | 882 | 67 | |
BC117074.1 | - | 1 | 660 | 40126393 | 67 | |
BC120777.1 | - | 1 | 660 | 40129700 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
