Assay Details
Gene Symbol: Cpsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018813.3 | NP_061283.2 | 14 - 15 | 1896 | 64 | |
GenBank mRNA | AK004317.1 | - | 1 - 2 | 106 | 64 | |
AK010566.1 | - | 7 - 8 | 963 | 64 | ||
AK045238.1 | - | 3 - 4 | 266 | 64 | ||
AK149953.1 | - | 14 - 15 | 1823 | 64 | ||
AK157205.1 | - | 14 - 15 | 1838 | 64 | ||
AK158036.1 | - | 14 - 15 | 1809 | 64 | ||
AK170842.1 | - | 14 - 15 | 1781 | 64 | ||
AK171838.1 | - | 14 - 15 | 1827 | 64 | ||
BC006748.1 | - | 9 - 10 | 1451 | 64 | ||
BC023297.1 | - | 14 - 15 | 1782 | 4192224 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
