Assay Details
Gene Symbol: Eya1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252192.1 | NP_001239121.1 | 13 - 14 | 1842 | 85 | |
NM_010164.2 | NP_034294.2 | 14 - 15 | 1958 | 85 | ||
GenBank mRNA | AJ007995.1 | - | 1 - 2 | 72 | 85 | |
AK042412.1 | - | 13 - 14 | 1765 | 85 | ||
AK153788.1 | - | 14 - 15 | 1964 | 85 | ||
AK162111.1 | - | 6 - 7 | 699 | 85 | ||
BC060260.1 | - | 14 - 15 | 1958 | 6848408 | 85 | |
BC066860.1 | - | 14 - 15 | 1612 | 30089692 | 85 | |
U61110.1 | - | 13 - 14 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
