Assay Details
Gene Symbol: Copb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015827.2 | NP_056642.1 | 1 - 2 | 90 | 87 | |
GenBank mRNA | AK088064.1 | - | 1 - 2 | 91 | 87 | |
AK153352.1 | - | 1 - 2 | 91 | 87 | ||
BC006675.1 | - | 1 - 2 | 77 | 3499585 | 87 | |
BC010842.1 | - | 1 - 2 | 80 | 87 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK088064.1, AK153352.1, BC006675.1, NM_015827.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
