Assay Details
Gene Symbol: Sgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013731.3 | NP_038759.1 | 1 - 2 | 114 | 76 | |
GenBank mRNA | AF169033.1 | - | 1 - 2 | 49 | 76 | |
AI747981.1 | - | 1 - 2 | 265 | 76 | ||
AK136559.1 | - | 1 - 2 | 66 | 76 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF169033.1, AK136559.1, NM_013731.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
