Assay Details
Gene Symbol: L3mbtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081338.1 | NP_001074807.1 | 18 - 19 | 2128 | 72 | |
GenBank mRNA | AK049859.1 | - | 17 - 18 | 2508 | 72 | |
AK220350.1 | - | 16 - 17 | 1876 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
