Assay Details
Gene Symbol: Dnmt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003960.4 | NP_001003960.2 | 18 - 19 | 2367 | 68 | |
NM_001003961.4 | NP_001003961.2 | 19 - 20 | 2427 | 68 | ||
NM_001003963.4 | NP_001003963.2 | 19 - 20 | 2427 | 68 | ||
NM_001122997.2 | NP_001116469.1 | 18 - 19 | 2315 | 68 | ||
NM_001271744.1 | NP_001258673.1 | 18 - 19 | 2312 | 68 | ||
NM_001271745.1 | NP_001258674.1 | 17 - 18 | 2252 | 68 | ||
NM_001271746.1 | NP_001258675.1 | 18 - 19 | 2312 | 68 | ||
NM_001271747.1 | NP_001258676.1 | 17 - 18 | 2252 | 68 | ||
NM_010068.5 | NP_034198.3 | 18 - 19 | 2367 | 68 | ||
GenBank mRNA | AF068626.2 | - | 19 - 20 | 2282 | 68 | |
AF068627.2 | - | 18 - 19 | 2222 | 68 | ||
AF068628.2 | - | 18 - 19 | 2222 | 68 | ||
AF151969.1 | - | 19 - 20 | 2425 | 68 | ||
AF151970.1 | - | 18 - 19 | 2365 | 68 | ||
AF151971.1 | - | 18 - 19 | 2365 | 68 | ||
AF151972.1 | - | 19 - 20 | 2425 | 68 | ||
AF151973.1 | - | 18 - 19 | 2310 | 68 | ||
AF151974.1 | - | 17 - 18 | 2250 | 68 | ||
AF151975.1 | - | 18 - 19 | 2310 | 68 | ||
AF151976.1 | - | 17 - 18 | 2250 | 68 | ||
AY078427.1 | - | 19 - 20 | 2282 | 68 | ||
BC105677.1 | - | 18 - 19 | 2147 | 40048574 | 68 | |
BC105922.1 | - | 18 - 19 | 2147 | 40048584 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
