Assay Details
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133847.3 | NP_598608.2 | 14 - 15 | 1766 | 68 | |
GenBank mRNA | AB093220.2 | - | 13 - 14 | 1510 | 68 | |
AK035622.1 | - | 14 - 15 | 1728 | 68 | ||
AK048158.1 | - | 14 - 15 | 1524 | 68 | ||
AK083541.1 | - | 14 - 15 | 1766 | 68 | ||
AK087048.1 | - | 14 - 15 | 1519 | 68 | ||
AK088012.1 | - | 14 - 15 | 1522 | 68 | ||
AK162435.1 | - | 14 - 15 | 1527 | 68 | ||
AK171487.1 | - | 14 - 15 | 1524 | 68 | ||
BC071208.1 | - | 1 - 2 | 585 | 68 | ||
BC110309.1 | - | 14 - 15 | 1522 | 5324218 | 68 | |
BC116320.2 | - | 14 - 15 | 1718 | 40096002 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
